ContactUsLinkComponent
Infant Heel Warmer
The site is currently experiencing technical difficulties. Please retry later. We apologize for any inconvenience.
Infant Heel Warmer
Catalog # 77591-830
New Product
undefined
Infant Heel Warmer
Catalog # 77591-830
Supplier Number:  ES23530
New Product
Specifications
  • Description:
    Infant heel warmers, non sterile
  • Environmentally Preferable:
  • Cat. No.:
    77591-830
Specifications
About this item
Infant heel warmers are designed to safely and comfortably increase blood flow to the infant's heel area. By increasing blood, the potential for painful bruising, infection, or cartilage damage caused by frequent or repeated heel sticks is minimized.
  • Designed and tested for infant safety
  • Conforms to infant's foot for even heat distribution
  • Trigger disk allows for quick and easy activation
  • Product is latex-free
A trigger disk allows for quick and easy activation. Heel warmers quickly and gently reach the optimal desired temperature of 105 °F.