HOBO® AC Current Switch Sensor
Catalog # 470346-426
Supplier: ONSET COMPUTER CORPORATION SE
Specifications
- Description:AC current switch sensor
- Cat. No.:470346-426
Specifications
About this item
Measures equipment runtime (time-of-use).
- Installs around a current-carrying wire to provide a relay signal indicating equipment on/off
- Adjustable threshold provides maximum application flexibility
- UL listed
Provides an adjustable current threshold of 0.5 to 175 A. Simply install the sensor around a current-carrying wire to the equipment to be monitored and then connect the sensor to a compatible data logger.
Ordering information: Compatible with HOBO UX90 and UX120-017 data loggers.