HOBO® No-Lead Water Flow Meter Sensor
470346-448
- Description:No-lead water flow meter sensor
- Cat. No.:470346-448
- Range:25 - 22.0
A compact, affordable water meter to measure cold and hot water flow rates.
- Measures cold and hot water flow rates
- Suitable for use in apartments, assisted-living communities, and other types of commercial buildings
- Configured for use with 3/4" pipes; use step-downs for smaller pipe
Designed for deployment in apartments, assisted living communities, and other commercial buildings.