HOBO® Wireless Temperature Sensor
Supplier: ONSET COMPUTER CORPORATION SE
Certificates
About this item
A battery-powered wireless sensor that to monitor air temperature.
- Measurement range of −40 to 100 °C (–40 to 212 °F)
- High accuracy of ±0.2 °C (±0.36 °F)
- Stainless-steel sensor tip
A cost-effective and scalable solution for web-enabled monitoring of air temperature. HOBOnet wireless sensors communicate data directly to a HOBO RX Station or pass data through other wireless sensors back to the central station. They are preconfigured and ready to deploy, and data is accessed through HOBOlink, Onset's innovative cloud software platform.
Caution: Includes lithium batteries.
Frequently Bought Together