HOBO® Water Level Data Logger Deluxe Kit
Ideal for monitoring water levels and temperatures in wells, streams, lakes and freshwater wetlands, as well as barometric pressure.
- Lightning protection; no long signal wires, and electronics are shielded in stainless-steel housing
- HOBOware Pro software provides easy conversion to accurate water level reading, fully compensated for barometric pressure, temperature, and water density
- Multiple-rate sampling allows faster sampling at critical times such as when pumping starts or stops
- Available in four depth ranges
- Ideal for use in wells, streams, lakes, wetlands, and tidal areas
- No-vent-tube design for easy reliable deployment
- Durable ceramic pressure sensor
Combining high accuracy water level measurements with out-of-the-box convenience, this money-saving kit includes everything needed to start logging right away.
: Bundle includes water depth logger (options of 30' or 100'), data logger for barometric pressure, waterproof shuttle with 5 interchangeable couplers, carrying case, and HOBO pro software.
Frequently Bought Together