HOBO® Water Level Sensor
470323-366
- Description:Water level sensor
- Cat. No.:470323-366
- Range:0 - 207 kPa (0 - 30 psia); approx. 0 - 9 m (0 - 30') of water depth at sea level, or 0 - 12 m (0 to 40') of water at 3,000 m (10,000') of altitude
Research grade level measurements and data monitoring for water pressure, temperature, and water level measurements.
- Non-vented design reduces maintenance
- Durable, ceramic sensor
- 3-point NIST-traceable calibration certificate
- Maximum error: 0.1% FS, 1.0 cm (0.03') water
These sensors are available in four depth ranges, and come in stainless-steel or titanium (recommended for use in saltwater) models.
: For use with HOBO MX2001 loggers, MicroRX Water Level Stations, RX3000 stations, and the HOBOnet Remote Water Level Monitoring System.