Order Entry
United States
ContactUsLinkComponent
CD437 ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
CD437 ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 82603-578
Supplier:  MilliporeSigma
CAS Number:  
CD437 ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 82603-578
Supplier:  MilliporeSigma
Supplier Number:  178496-5MG
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Size:
    5 mg
  • Cat. No.:
    82603-578
  • Supplier no.:
    178496-5MG

Specifications

About this item

 :
 :398.4936 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:125316-60-1
Formula:C₂₇H₂₆O₃
MDL Number:MFCD03106506