D,L-threo-PDMP hydrochloride ≥98% (by HPLC), Calbiochem®, Sigma-Aldrich®
Catalog # 80053-612
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampule
- Size:50 mg
- Cat. No.:80053-612
- Supplier no.:513100-50MG
Specifications
About this item
:
:390.6·36.5 g/mol
Storage Temperature:Freezer
CAS Number:80938-69-8
Formula:C₂₃H₃₈N₂O₃·HCl
MDL Number:MFCD00236490