Microvette® with Capillary
Supplier: SARSTEDT INC
New Product
Certificates
About this item
Capillary blood collection facilitates diagnostics with a small volume of blood.
- Closure: Cap
- Outer container with skirted base and graduations
- End-to-end capillary pre-assembled
- Smooth cap - special lid design reduces the aerosol effect
- Conical inner tube: After centrifugation, higher supernatant for easier collection of plasma - cylindrical inner tube: Optimal tube shape ensures the sample is well mixed
- Two collection techniques: Capillary or collection rim
The capillary blood collection systems are among the most versatile on the market. Microvette® with nominal volumes between 100 and 500 μl, conical or cylindrical inner shapes, as well as your choice of collection technique - end-to-end or with a collection rim.
Frequently Bought Together