Orders LinkContactUsLinkComponent
Feces Tubes with Flat Base and Push Cap
Feces Tubes with Flat Base and Push Cap
Supplier:   SARSTEDT INC
Feces Tubes with Flat Base and Push Cap
Supplier:  SARSTEDT INC
Certificates
Feces Tubes with Flat Base and Push Cap
Feces Tubes with Flat Base and Push Cap
2 Options Available Below
Product Details & Documents
In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, this offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
  • Material: PS
  • Particularly liquid and odor-tight
  • Larger opening to better collect the sample
  • Stands firmly due to skirted base
To hygienically collect stool samples, all collection vessels are fitted with integral spoons. Fecal collection tubes combined with a broad and sturdy scoop in the screw cap make sampling easy.
Product Family Options
Product Information
  • Material
    Sterility
    Ø
    Length
    Packaging
    Availability
    Price
  • PS
    Non sterile
    23.5 mm
    75 mm
    BAGA
    Log in for availability
    Log in for availability
    Case of 500
    Case of 500 - $227.19
    $227.19
    Case of 500
       $227.19
    Supplier Number: 80.620
  • PS
    Sterile
    23.5 mm
    75 mm
    BAGA
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $35.45
    $35.45
    Log in for availability
    Case of 250
    Case of 250 - $177.25
    $177.25
    Pack of 50
       $35.45
    Case of 250
       $177.25
    Supplier Number: 80.621