Order Entry
United States
ContactUsLinkComponent
Endo-β-galactosidase from Bacteroides fragilis recombinant expressed in E. coli, Calbiochem®, Sigma-Aldrich®
Endo-β-galactosidase from Bacteroides fragilis recombinant expressed in E. coli, Calbiochem®, Sigma-Aldrich®
Catalog # 82603-556
Supplier:  MilliporeSigma
CAS Number:  
Endo-β-galactosidase from Bacteroides fragilis recombinant expressed in E. coli, Calbiochem®, Sigma-Aldrich®
Catalog # 82603-556
Supplier:  MilliporeSigma
Supplier Number:  345811-50UL
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Cat. No.:
    82603-556
  • Supplier no.:
    345811-50UL

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
MDL Number:MFCD00131433