DSP (Dithiobis(succinimidyl propionate)) ≥94% (by HPLC), water soluble, Calbiochem®, Sigma-Aldrich®
Catalog # EM322133-100MG
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Size:100 mg
- Cat. No.:EM322133-100MG
- Supplier no.:322133-100MG
Specifications
About this item
:
:404.41 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:57757-57-0
Formula:C₁₄H₁₆N₂O₈S₂
MDL Number:MFCD00042045