Orders LinkContactUsLinkComponent
Screw Cap Tubes with Skirted Conical Base, PS
Screw Cap Tubes with Skirted Conical Base, PS
Supplier:   SARSTEDT INC
Screw Cap Tubes with Skirted Conical Base, PS
Supplier:  SARSTEDT INC
Certificates
Screw Cap Tubes with Skirted Conical Base, PS
Screw Cap Tubes with Skirted Conical Base, PS
3 Options Available Below
Product Details & Documents
High-quality, safe and robust containers are essential for mixing, processing and storing samples. As one of the world’s largest manufacturers of tubes and storage containers, this range offers the right product for every laboratory and analytical requirement.
  • Material: PS
  • Graduations: No
  • Base shape: Skirted conical base
  • Corresponds to the requirement: ADR
  • Tube with skirted base ensures that it stands securely
  • Polystyrene (PS) tubes for maximum transparency
  • Especially secure cap
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base; with or without skirted base; with stopper or screw cap; with or without print or labels.
Product Family Options
Product Information
  • Color
    Volume
    Ø
    Length
    Cap included
    Cap color
    Cap material
    Cap type
    Max. rcf
    Availability
    Price
  • 77587-832
    Screw cap tubes with skirted conical base, cap enclosed
    Screw cap tubes with skirted conical base, cap enclosed
    Transparent
    10 ml
    16 mm
    97 mm
    Yes
    Natural
    HDPE
    Screw cap
    4000×g
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $148.66
    $148.66
    Log in for availability
    Case of 1000
    Case of 1000 - $297.32
    $297.32
    Pack of 500
       $148.66
    Case of 1000
       $297.32
    Supplier Number: 60.9921.821
  • 77587-834
    Screw cap tubes with skirted conical base, cap assembled
    Screw cap tubes with skirted conical base, cap assembled
    Transparent
    10 ml
    16 mm
    97 mm
    Yes
    Yellow
    HDPE
    Screw cap
    4000×g
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $30.53
    $30.53
    Log in for availability
    Case of 1000
    Case of 1000 - $305.30
    $305.30
    Pack of 100
       $30.53
    Case of 1000
       $305.30
    Supplier Number: 60.9921.829
  • 77587-836
    Screw cap tubes with skirted conical base, cap assembled
    Screw cap tubes with skirted conical base, cap assembled
    Transparent
    10 ml
    16 mm
    97 mm
    Yes
    Yellow
    HDPE
    Screw cap
    4000×g
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $32.96
    $32.96
    Log in for availability
    Case of 1000
    Case of 1000 - $329.60
    $329.60
    Pack of 100
       $32.96
    Case of 1000
       $329.60
    Supplier Number: 60.9921.830