Orders LinkContactUsLinkComponent
Micro Needles
Micro Needles
Supplier:   SARSTEDT INC
Micro Needles
Supplier:  SARSTEDT INC
Certificates
Micro Needles
3 Options Available Below
Product Details & Documents
The micro needles were developed to meet the specific needs of neonatology. It has a grooved holder to ensure safe puncture and enables turning the needle in the vein for better positioning. The short design of the needle means a minimal dead volume during sample collection.
  • Ideal for venous blood collection of even the tiniest quantities
  • Minimal blood loss with no residual volume in the holder
  • Ridged holder for a secure grip allows optimal handling of the needle
  • Can be rotated 360° in difficult vein conditions
  • Exceptional needle quality and exposed outlet opening enable free blood flow
These pediatric blood collection products make the process of collecting blood from your smallest patients as gentle and painless as possible. The priority is minimizing the fill volume of each collection and keeping the diagnostic blood loss as low as possible.
Documents
Product Family Options
Product Information
  • Length
    Sterility
    Availability
    Price
  • 77589-490
    Micro needle 21G × ¾", green
    Micro needle 21G × ¾", green
    36 mm
    Sterile, non-pyrogenic/endotoxin-free
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $147.87
    $147.87
    Log in for availability
    Case of 500
    Case of 500 - $1,478.70
    $1,478.70
    Pack of 50
       $147.87
    Case of 500
       $1,478.70
    Supplier Number: 85.921
    Specifications:
  • 77589-492
    Micro needle 23G × ¾", blue
    Micro needle 23G × ¾", blue
    36 mm
    Sterile, non-pyrogenic/endotoxin-free
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $147.87
    $147.87
    Log in for availability
    Case of 500
    Case of 500 - $1,478.70
    $1,478.70
    Pack of 50
       $147.87
    Case of 500
       $1,478.70
    Supplier Number: 85.923
    Specifications:
  • 77589-494
    Micro needle 25G × ¾", orange
    Micro needle 25G × ¾", orange
    36 mm
    Sterile, non-pyrogenic/endotoxin-free
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $147.87
    $147.87
    Log in for availability
    Case of 500
    Case of 500 - $1,478.70
    $1,478.70
    Pack of 50
       $147.87
    Case of 500
       $1,478.70
    Supplier Number: 85.925
    Specifications:
Recommendations