Sample Tubes for Special Analyzers
Supplier: SARSTEDT INC
New Product
Certificates
About this item
High-quality, safe and robust containers are essential for mixing, processing and storing samples. Sarstedt offers the right product for every laboratory and analytical requirement.
- Graduations: No
- Conical base shape for optimal sample yield
- Polypropylene (PP) tubes for thermal, mechanical and chemical applications
This broad product portfolio in the field of reagent and centrifuge tubes includes options with round, conical or flat base, with or without skirted base, with stopper or screw cap, with or without print or labels.
Frequently Bought Together