ContactUsLinkComponent
G.S.C. International Bell and Vacuum Demonstration Kit
G.S.C. International Bell and Vacuum Demonstration Kit
  470356-992
New Product
undefined
G.S.C. International Bell and Vacuum Demonstration Kit
  470356-992
 :  BELLKT
New Product
  • Description:
    Bell and vacuum physics demonstration kit
  • Cat. No.:
    470356-992
Bell and vacuum kit for classroom and homeschools is designed to enhance science education by providing a tangible, interactive learning experience for students. It's perfect for demonstrating complex physics concepts in a clear and engaging way, making it an ideal addition to any school classroom or science lab.
  • This kit aligns with educational standards for physics and science, providing a practical demonstration of acoustic properties and the nature of sound
  • An excellent resource for teachers aiming to reinforce key concepts and enhance student understanding through practical, hands-on learning experiences
The kit allows students to explore the principles of sound propagation by contrasting how sound travels through air versus a vacuum. Through hands-on experimentation, students can witness firsthand how sound waves diminish significantly in a vacuum, deepening their understanding of this fundamental scientific concept.
The kit comes with all the necessary components, including an airtight jar with a 4.75" diameter, an air gauge for monitoring air displacement, a user-friendly hand pump, a buzzer, and a hose. This comprehensive setup ensures that educators can easily integrate the kit into their lesson plans without needing additional equipment.
By using this kit, students can engage in memorable experiments that bring physics to life. It promotes active learning and encourages curiosity and critical thinking, helping students to better retain information and apply their knowledge to real-world scenarios.
 : This product can exposes user to chemicals including styrene and lead. Contains latex, which can cause allergic reactions in some people. Use in a laboratory or educational setting only. Not for children 13 and under.