Orders LinkContactUsLinkComponent
Absorbent Liners for Transport Containers
Absorbent Liners for Transport Containers
Supplier:   SARSTEDT INC
Absorbent Liners for Transport Containers
Supplier:  SARSTEDT INC
Certificates
Absorbent Liners for Transport Containers
Absorbent Liners for Transport Containers
3 Options Available Below
Product Details & Documents
SARSTEDT offers sample transport systems and secondary containers for a wide variety of transport requirements.
  • Material: Cellulose
  • Separate absorbent liners suitable for various mailing containers
  • Various designs for different absorption capacities
For use with mailing containers with screw cap; a secure container for blood, urine and stool sample tubes.
Product Family Options
Product Information
  • Dimensions
    Packaging
    Availability
    Price
  • 77589-104
    Absorbent liners for mailing container 84×30 mm, 8 ml suction capacity
    Absorbent liners for mailing container 84×30 mm, 8 ml suction capacity
    75×65 mm
    CS
    Log in for availability
    Log in for availability
    Case of 2,000
    Case of 2,000 - $295.75
    $295.75
    Case of 2,000
       $295.75
    Supplier Number: 78.684
  • 77589-100
    Absorbent liners for mailing container 126ר 30 mm, 12 ml suction capacity
    Absorbent liners for mailing container 126ר 30 mm, 12 ml suction capacity
    75×90 mm
    CS
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $178.04
    $178.04
    Log in for availability
    Case of 4000
    Case of 4000 - $712.16
    $712.16
    Pack of 1,000
       $178.04
    Case of 4000
       $712.16
    Supplier Number: 78.670
  • 77589-102
    Absorbent liners for transport containers, 10 ml suction capacity
    Absorbent liners for transport containers, 10 ml suction capacity
    80×55 mm
    CS
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $47.45
    $47.45
    Log in for availability
    Case of 2500
    Case of 2500 - $1,186.25
    $1,186.25
    Pack of 100
       $47.45
    Case of 2500
       $1,186.25
    Supplier Number: 78.671