Anti-NP-COV Mouse Antibody (Biotin)
77585-254
New Product
- Antigen Symbol:NP-COV
- Clonality:Monoclonal
- Conjugation:Biotin
- Host:Mouse
- Reactivity:SARS-CoV
- Size:20 µg
- Cat. No.:77585-254
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS-CoV Nucleoprotein / NP (Catalog#40143-V08B; NP_828858.1; Met1-Ala422) and then biotinylated. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and then biotinylated.