Feces Tubes
About this item
To meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, Sarstedt offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
- Material: PP
- Particularly liquid and odor-tight
- Larger opening to better collect the sample
- Secure screw cap for sending samples
- Stands firmly due to skirted base
- Options with paper labels and white material for light protection
To hygienically collect stool samples, all collection vessels are fitted with integral spoons. Fecal collection tubes combined with a broad and sturdy scoop in the screw cap make sampling easy.
9 Options Available Below
Product Details & Documents
To meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, Sarstedt offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
- Material: PP
- Particularly liquid and odor-tight
- Larger opening to better collect the sample
- Secure screw cap for sending samples
- Stands firmly due to skirted base
- Options with paper labels and white material for light protection
To hygienically collect stool samples, all collection vessels are fitted with integral spoons. Fecal collection tubes combined with a broad and sturdy scoop in the screw cap make sampling easy.
Product Family Options
Product Information
- MaterialSterilityColorØLengthPackagingAvailabilityPrice
- 77589-122Feces tubesSupplier Number: 80.623
- 77589-126Feces tubesSupplier Number: 80.623.111
- 77589-140Feces tubesSupplier Number: 80.734
- 77589-142Feces tubesSupplier Number: 80.734.001
- 77589-144Feces tubes, with paper labelFeces tubes, with paper labelPPNon sterileTransparent20 mm76 mmBAGALog in for availabilityCase of 500$254.62Supplier Number: 80.734.301
- 77589-146Feces tubes, with paper labelFeces tubes, with paper labelPPSterileTransparent20 mm76 mmBAGALog in for availabilityCase of 500$323.91Supplier Number: 80.734.311
- 77589-148Feces tubes, with paper labelFeces tubes, with paper labelPP, whiteNon sterileTransparent20 mm76 mmBAGALog in for availabilityCase of 500$228.62Supplier Number: 80.734.401
- 77589-118Feces tubesFeces tubesPPSterileTransparent25 mm107 mmBAGAPack of 50$42.85Case of 400$342.80Supplier Number: 80.622
- 77589-120Feces tubesSupplier Number: 80.622.111