ReadyTag Tag Anti-HA Mouse Antibody [clone: 12CA5]
Catalog # 77580-506
Supplier: Bio X Cell
New Product
Specifications
- Antibody Type:Primary
- Antigen Symbol:HA
- Clone:12CA5
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Western Blot:Yes
- Size:5 mg
- Antibody Fragment:Tag
- Tested Applications:ELISA
- Cat. No.:77580-506
Specifications
About this item
Type: Primary
Antigen: HA
Clonality:
Clone: 12CA5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: