RecombiMAb Anti-CTLA-4 Mouse Recombinant Antibody [clone: 9H10-CP146]
New Product
Type: Primary
Antigen: CTLA-4
Clonality: Recombinant
Clone: 9H10-CP146
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
- Catalog No:
- 77580-218
- 77580-216
- 77580-214
- 77580-212
- 77580-210
- Antigen Symbol:
- CTLA-4
- CTLA-4
- CTLA-4
- CTLA-4
- CTLA-4
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Clone:
- 9H10-CP146
- 9H10-CP146
- 9H10-CP146
- 9H10-CP146
- 9H10-CP146
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Western Blot:
- Yes
- Yes
- Yes
- Yes
- Yes
- Tested Applications:
- in vivo CTLA-4 neutralization*, in vitro CTLA-4 neutralization*
*Reported for the original Syrian hamster IgG 9H10 antibody - in vivo CTLA-4 neutralization*, in vitro CTLA-4 neutralization*
*Reported for the original Syrian hamster IgG 9H10 antibody - in vivo CTLA-4 neutralization*, in vitro CTLA-4 neutralization*
*Reported for the original Syrian hamster IgG 9H10 antibody - in vivo CTLA-4 neutralization*, in vitro CTLA-4 neutralization*
*Reported for the original Syrian hamster IgG 9H10 antibody - in vivo CTLA-4 neutralization*, in vitro CTLA-4 neutralization*
*Reported for the original Syrian hamster IgG 9H10 antibody
Frequently Bought Together