RecombiMAb Anti-4-1BBL Mouse Recombinant Antibody [clone: TKS-1-CP039]
Catalog # 77580-082
Supplier: Bio X Cell
New Product
Specifications
- Antibody Type:Primary
- Antigen Symbol:4-1BBL
- Clonality:Recombinant
- Clone:TKS-1-CP039
- ELISA:Yes
- Host:Mouse
- Reactivity:Mouse
- Size:1 mg
- Tested Applications:in vivo 4-1BBL blockade*
*Reported for the original rat IgG2a TKS-1 antibody - Cat. No.:77580-082
Specifications
About this item
Type: Primary
Antigen: 4-1BBL
Clonality: Recombinant
Clone: TKS-1-CP039
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse