RecombiMAb Anti-CD40L Mouse Recombinant Antibody [clone: MR-1-CP032]
Catalog # 77580-074
Supplier: Bio X Cell
New Product
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD40L
- Clonality:Recombinant
- Clone:MR-1-CP032
- Host:Mouse
- Western Blot:Yes
- Size:1 mg
- Tested Applications:in vivo blocking of CD40/CD40L signaling*, in vitro blocking of CD40/CD40L signaling*
*Reported for the original Armenian hamster IgG MR-1 antibody - Cat. No.:77580-074
Specifications
About this item
Type: Primary
Antigen: CD40L
Clonality: Recombinant
Clone: MR-1-CP032
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse