RecombiMAb Anti-PD-1 Mouse Recombinant Antibody [clone: 29F.1A12™-CP004]
77580-044
New Product
- Antibody Type:Primary
- Antigen Symbol:PD-1
- Clonality:Recombinant
- Clone:29F.1A12™-CP004
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Western Blot:Yes
- Size:1 mg
- Tested Applications:in vivo blocking of PD-1/PD-L signaling*, in vitro PD-1 neutralization*, Immunohistochemistry (frozen)*,
*Reported for the original rat IgG2a 29F.1A12 antibody - Cat. No.:77580-044
Type: Primary
Antigen: PD-1
Clonality: Recombinant
Clone: 29F.1A12™-CP004
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse