InVivoMAb Rat IgG2b Isotype Control Anti-Trinitrophenol-Keyhole Limpet Hemocyanin
New Product
Type: Control
Antigen: IgG2b
Clonality: Monoclonal
Clone: TNP-4G1
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
- Catalog No:
- 77579-798
- 77579-796
- 77579-794
- 77579-792
- 77579-790
- Antigen Symbol:
- IgG2b
- IgG2b
- IgG2b
- IgG2b
- IgG2b
- Antibody Type:
- Control
- Control
- Control
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- TNP-4G1
- TNP-4G1
- TNP-4G1
- TNP-4G1
- TNP-4G1
Frequently Bought Together