InVivoMAb Anti-MAGE-C2 Mouse Monoclonal Antibody [clone: LX-CT10.5]
Supplier: Bio X Cell
New Product
Certificates
About this item
Type: Primary
Antigen: MAGE-C2
Clonality: Monoclonal
Clone: LX-CT10.5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- 77578-908
- 77578-906
- 77578-904
- 77578-902
- 77578-900
- Antigen Symbol:
- MAGE-C2
- MAGE-C2
- MAGE-C2
- MAGE-C2
- MAGE-C2
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- LX-CT10.5
- LX-CT10.5
- LX-CT10.5
- LX-CT10.5
- LX-CT10.5
- Reactivity:
- Human
- Human
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Western Blot:
- Yes
- Yes
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Yes
- Yes
- Tested Applications:
- Immunohistochemistry (paraffin)
- Immunohistochemistry (paraffin)
- Immunohistochemistry (paraffin)
- Immunohistochemistry (paraffin)
- Immunohistochemistry (paraffin)
Specifications
Frequently Bought Together