Faeces Containers
About this item
In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, SARSTEDT offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
- Material: PP
- Label: With paper label
- Particularly liquid and odor-tight
- Larger opening to better collect the sample
- Secure screw cap for sending samples
- Stands firmly due to skirted base
Fecal containers with large opening for hygienic collection.
2 Options Available Below
Product Details & Documents
In order to meet the various requirements of hygienic collection, safe transportation and storage of diagnostic sample materials, SARSTEDT offers many high-quality products for collecting urine, stool, saliva, CSF and tissue samples.
- Material: PP
- Label: With paper label
- Particularly liquid and odor-tight
- Larger opening to better collect the sample
- Secure screw cap for sending samples
- Stands firmly due to skirted base
Fecal containers with large opening for hygienic collection.
Product Family Options
Product Information
- MaterialColorØLengthCap colorClosure typePackagingAvailabilityPrice
- 77589-150Feces tube 54ר 28 mmSupplier Number: 80.9924.014
- 77589-152Feces tube 55ר 44 mmFeces tube 55ר 44 mmPPWhite44 mm55 mmBrownScrew cap250/bag, 500/caseLog in for availabilityCase of 500$273.81Supplier Number: 80.9924.027