Orders LinkContactUsLinkComponent
Safety Lancets
Safety Lancets
Supplier:   SARSTEDT INC
Safety Lancets
Supplier:  SARSTEDT INC
Certificates
Safety Lancets
5 Options Available Below
Product Details & Documents
Sarstedt safety lancets guarantee optimal safety for patients and users and are flexible with regard to penetration depth and required quantity of blood. The needle and blade are always safely concealed in the lancet body before and after use, which prevents needlestick injuries and cross-contamination.
  • Safety for users and patients
  • Sterile, single-use system
  • Patient-friendly handling activated by a release button
  • Large selection of penetration depths and widths depending on the quantity of blood and skin conditions
Sarstedt's pre-analytical product range includes products for optimal specimen collection. The capillary collection systems are among the most versatile on the market.
Product Family Options
Product Information
  • Gauge
    Color
    Availability
    Price
  • 77589-426
    Safety lancet, penetration depth: 1.2 mm, blade width: 1.5 mm
    Safety lancet, penetration depth: 1.2 mm, blade width: 1.5 mm
    Pink
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 200
    Pack of 200 - $87.57
    $87.57
    Log in for availability
    Case of 1200
    Case of 1200 - $525.42
    $525.42
    Pack of 200
       $87.57
    Case of 1200
       $525.42
    Supplier Number: 85.1019
    Specifications:
  • 77589-418
    Safety lancet, penetration depth: 1.6 mm
    Safety lancet, penetration depth: 1.6 mm
    28 G
    Light blue
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 200
    Pack of 200 - $71.66
    $71.66
    Log in for availability
    Case of 1200
    Case of 1200 - $429.96
    $429.96
    Pack of 200
       $71.66
    Case of 1200
       $429.96
    Supplier Number: 85.1015
    Specifications:
  • 77589-424
    Safety lancet, penetration depth: 1.6 mm, blade width: 1.5 mm
    Safety lancet, penetration depth: 1.6 mm, blade width: 1.5 mm
    Violet
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 200
    Pack of 200 - $87.57
    $87.57
    Log in for availability
    Case of 1200
    Case of 1200 - $525.42
    $525.42
    Pack of 200
       $87.57
    Case of 1200
       $525.42
    Supplier Number: 85.1018
    Specifications:
  • 77589-420
    Safety lancet, penetration depth: 1.8 mm
    Safety lancet, penetration depth: 1.8 mm
    21 G
    Green
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 200
    Pack of 200 - $71.66
    $71.66
    Log in for availability
    Case of 1200
    Case of 1200 - $429.96
    $429.96
    Pack of 200
       $71.66
    Case of 1200
       $429.96
    Supplier Number: 85.1016
    Specifications:
  • 77589-422
    Safety lancet, penetration depth: 1.8 mm
    Safety lancet, penetration depth: 1.8 mm
    18 G
    Yellow
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 200
    Pack of 200 - $71.66
    $71.66
    Log in for availability
    Case of 1200
    Case of 1200 - $429.96
    $429.96
    Pack of 200
       $71.66
    Case of 1200
       $429.96
    Supplier Number: 85.1017
    Specifications: