Prepared Micro Sample Tubes with Screw Cap and Label
Supplier: SARSTEDT INC
New Product
Certificates
About this item
The prepared micro tubes are available with push cap closure or screw cap with integral O-ring.
- Base: Skirted conical base
- Ideal collection vessel for small volumes of blood
- Screw cap with O-ring ideal for transport and storage
- Screw cap with pierceable membrane available for analyzer adaptation
- Writable paper label with graduations to check fill level
- Ø: 10.8 mm
- Length without/with cap: 45/47 mm
These micro volume tubes are prepared with standard additives for versatile low-volume requirements.
Frequently Bought Together