RecombiMAb Mouse IgG2a (LALA-PG) Isotype control Unknown Specificity
New Product
Type: Control
Antigen: IgG2a
Clonality: Recombinant
Clone: MOPC-21-CP156
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
- Catalog No:
- 77580-318
- 77580-316
- 77580-314
- 77580-312
- 77580-310
- Antigen Symbol:
- IgG2a
- IgG2a
- IgG2a
- IgG2a
- IgG2a
- Antibody Type:
- Control
- Control
- Control
- Control
- Control
- Clonality:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Clone:
- MOPC-21-CP156
- MOPC-21-CP156
- MOPC-21-CP156
- MOPC-21-CP156
- MOPC-21-CP156
Frequently Bought Together