RecombiMAb Human IgG4 (S228P) Isotype control Anti-Respiratory Syncytial Virus
Supplier: Bio X Cell
New Product
Certificates
About this item
Type: Control
Antigen: IgG4
Clonality: Recombinant
Clone: Palivizumab-CP152
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Specifications
- Catalog No:
- 77580-270
- 77580-278
- 77580-276
- 77580-274
- 77580-272
- Antigen Symbol:
- IgG4
- IgG4
- IgG4
- IgG4
- IgG4
- Antibody Type:
- Control
- Control
- Control
- Control
- Control
- Clonality:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Clone:
- Palivizumab-CP152
- Palivizumab-CP152
- Palivizumab-CP152
- Palivizumab-CP152
- Palivizumab-CP152
Specifications
Frequently Bought Together