InVivoPlus Mouse IgG2c Isotype control Anti-Dengue Virus
Supplier: Bio X Cell
New Product
Certificates
About this item
Type: Control
Antigen: IgG2c
Clonality:
Clone: DV5-1
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Dengue virus
Specifications
- Catalog No:
- 77580-038
- 77580-036
- 77580-034
- 77580-032
- Antigen Symbol:
- IgG2c
- IgG2c
- IgG2c
- IgG2c
- Antibody Type:
- Control
- Control
- Control
- Control
- Clone:
- DV5-1
- DV5-1
- DV5-1
- DV5-1
- Reactivity:
- Dengue virus
- Dengue virus
- Dengue virus
- Dengue virus
Specifications
Frequently Bought Together