RecombiMAb Anti-PD-1 Mouse Recombinant Antibody [clone: RMP1-14-CP157]
New Product
Type: Primary
Antigen: PD-1
Clonality: Recombinant
Clone: RMP1-14-CP157
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
- Catalog No:
- 77580-328
- 77580-326
- 77580-324
- 77580-322
- 77580-320
- Antigen Symbol:
- PD-1
- PD-1
- PD-1
- PD-1
- PD-1
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Clone:
- RMP1-14-CP157
- RMP1-14-CP157
- RMP1-14-CP157
- RMP1-14-CP157
- RMP1-14-CP157
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Tested Applications:
- in vivo blocking of PD-1/PD-L signaling* *Reported for the original rat IgG2a RMP1-14 antibody
- in vivo blocking of PD-1/PD-L signaling* *Reported for the original rat IgG2a RMP1-14 antibody
- in vivo blocking of PD-1/PD-L signaling* *Reported for the original rat IgG2a RMP1-14 antibody
- in vivo blocking of PD-1/PD-L signaling* *Reported for the original rat IgG2a RMP1-14 antibody
- in vivo blocking of PD-1/PD-L signaling* *Reported for the original rat IgG2a RMP1-14 antibody
Frequently Bought Together