Order Entry
United States
ContactUsLinkComponent
 
RecombiMAb Anti-PD-1 Mouse Recombinant Antibody [clone: RMP1-14-CP153]
New Product
 
RecombiMAb Anti-PD-1 Mouse Recombinant Antibody [clone: RMP1-14-CP153]
New Product

 

  • Catalog No:
  • 77580-288
  • 77580-286
  • 77580-284
  • 77580-282
  • 77580-280
  • Antigen Symbol:
  • PD-1
  • PD-1
  • PD-1
  • PD-1
  • PD-1
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Clone:
  • RMP1-14-CP153
  • RMP1-14-CP153
  • RMP1-14-CP153
  • RMP1-14-CP153
  • RMP1-14-CP153
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Tested Applications:
  • in vivo blocking of PD-1/PD-L signaling* *Reported for the original rat IgG2a RMP1-14 antibody
  • in vivo blocking of PD-1/PD-L signaling* *Reported for the original rat IgG2a RMP1-14 antibody
  • in vivo blocking of PD-1/PD-L signaling* *Reported for the original rat IgG2a RMP1-14 antibody
  • in vivo blocking of PD-1/PD-L signaling* *Reported for the original rat IgG2a RMP1-14 antibody
  • in vivo blocking of PD-1/PD-L signaling* *Reported for the original rat IgG2a RMP1-14 antibody

 

Frequently Bought Together