Order Entry
United States
ContactUsLinkComponent
RecombiMAb Anti-Ly6G/Ly6C Mouse Recombinant Antibody [clone: RB6-8C5-CP135]
RecombiMAb Anti-Ly6G/Ly6C Mouse Recombinant Antibody [clone: RB6-8C5-CP135]
Supplier:  Bio X Cell
New Product
Certificates
RecombiMAb Anti-Ly6G/Ly6C Mouse Recombinant Antibody [clone: RB6-8C5-CP135]
Supplier:  Bio X Cell
New Product

Specifications

  • Catalog No:
  • 77580-204
  • 77580-202
  • 77580-200
  • 77580-208
  • 77580-206
  • Antigen Symbol:
  • Ly6G/Ly6C
  • Ly6G/Ly6C
  • Ly6G/Ly6C
  • Ly6G/Ly6C
  • Ly6G/Ly6C
  • Antibody Type:
  • Primary
  • Primary
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Recombinant
  • Clone:
  • RB6-8C5-CP135
  • RB6-8C5-CP135
  • RB6-8C5-CP135
  • RB6-8C5-CP135
  • RB6-8C5-CP135
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Mouse
  • Flow Cytometry:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Tested Applications:
  • in vivo depletion of Gr-1+ myeloid cells**, Immunohistochemistry (paraffin)*, Immunohistochemistry (frozen)*, *Reported for the original rat IgG2b RB6-8C5 antibody
  • in vivo depletion of Gr-1+ myeloid cells**, Immunohistochemistry (paraffin)*, Immunohistochemistry (frozen)*, *Reported for the original rat IgG2b RB6-8C5 antibody
  • in vivo depletion of Gr-1+ myeloid cells**, Immunohistochemistry (paraffin)*, Immunohistochemistry (frozen)*, *Reported for the original rat IgG2b RB6-8C5 antibody
  • in vivo depletion of Gr-1+ myeloid cells**, Immunohistochemistry (paraffin)*, Immunohistochemistry (frozen)*, *Reported for the original rat IgG2b RB6-8C5 antibody
  • in vivo depletion of Gr-1+ myeloid cells**, Immunohistochemistry (paraffin)*, Immunohistochemistry (frozen)*, *Reported for the original rat IgG2b RB6-8C5 antibody

Specifications

Frequently Bought Together