RecombiMAb Anti-CD71 Mouse Recombinant Antibody [clone: R17]
New Product
Type: Primary
Antigen: CD71
Clonality: Recombinant
Clone: R17
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse
- Catalog No:
- 77580-158
- 77580-156
- 77580-154
- 77580-152
- 77580-150
- Antigen Symbol:
- CD71
- CD71
- CD71
- CD71
- CD71
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Clone:
- R17
- R17
- R17
- R17
- R17
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Tested Applications:
- in vivo depletion of CD71+ cells*, *Reported for the original rat IgG2a R17 217.1.3 antibody
- in vivo depletion of CD71+ cells*, *Reported for the original rat IgG2a R17 217.1.3 antibody
- in vivo depletion of CD71+ cells*, *Reported for the original rat IgG2a R17 217.1.3 antibody
- in vivo depletion of CD71+ cells*, *Reported for the original rat IgG2a R17 217.1.3 antibody
- in vivo depletion of CD71+ cells*, *Reported for the original rat IgG2a R17 217.1.3 antibody
Frequently Bought Together