RecombiMAb Anti-CD28 Mouse Recombinant Antibody [clone: D665-CP042]
Catalog # 77580-088
Supplier: Bio X Cell
New Product
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD28
- Clonality:Recombinant
- Clone:D665-CP042
- Host:Mouse
- Reactivity:Mouse
- Size:1 mg
- Tested Applications:in vivo T cell stimulation/activation*, in vitro T cell stimulation/activation* *Reported for the original mouse IgG1 D665 antibody
- Cat. No.:77580-088
Specifications
About this item
Type: Primary
Antigen: CD28
Clonality: Recombinant
Clone: D665-CP042
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse