RecombiMAb Anti-4-1BB Mouse Recombinant Antibody [clone: LOB12.3-CP038]
Catalog # 77580-080
Supplier: Bio X Cell
New Product
Specifications
- Antibody Type:Primary
- Antigen Symbol:4-1BB
- Clonality:Recombinant
- Clone:LOB12.3-CP038
- Host:Mouse
- Reactivity:Mouse
- Size:1 mg
- Tested Applications:in vivo activation of 4-1BB* *Reported for the original rat IgG1 LOB12.3 antibody
- Cat. No.:77580-080
Specifications
About this item
Type: Primary
Antigen: 4-1BB
Clonality: Recombinant
Clone: LOB12.3-CP038
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse