RecombiMAb Anti-GITR Mouse Recombinant Antibody [clone: DTA-1-CP029]
Catalog # 77580-070
Supplier: Bio X Cell
New Product
Specifications
- Antibody Type:Primary
- Antigen Symbol:GITR
- Clonality:Recombinant
- Clone:DTA-1-CP029
- Host:Mouse
- Size:1 mg
- Tested Applications:in vivo GITR stimulation* *Reported for the original rat IgG2b DTA-1 antibody
- Cat. No.:77580-070
Specifications
About this item
Type: Primary
Antigen: GITR
Clonality: Recombinant
Clone: DTA-1-CP029
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse