RecombiMAb Anti-CD16/CD32 Mouse Recombinant Antibody [clone: 2.4G2-CP026]
77580-064
New Product
- Antibody Type:Primary
- Antigen Symbol:CD16/CD32
- Clonality:Recombinant
- Clone:2.4G2-CP026
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Size:1 mg
- Tested Applications:in vivo Fc receptor blocking* Fc receptor blocking, flow cytometry* Fc receptor blocking, immunofluorescence**Reported for the original rat IgG2b 2.4G2 antibody
- Cat. No.:77580-064
Type: Primary
Antigen: CD16/CD32
Clonality: Recombinant
Clone: 2.4G2-CP026
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse