RecombiMAb Anti-LAG-3 Mouse Recombinant Antibody [clone: C9B7W-CP014]
Catalog # 77580-056
Supplier: Bio X Cell
New Product
Specifications
- Antibody Type:Primary
- Antigen Symbol:LAG-3
- Clonality:Recombinant
- Clone:C9B7W-CP014
- Flow Cytometry:Yes
- Host:Mouse
- Reactivity:Mouse
- Western Blot:Yes
- Size:1 mg
- Tested Applications:in vivo LAG-3 neutralization*, in vitro LAG-3 neutralization*** *Reported for the original rat IgG1 C9B7W antibody
- Cat. No.:77580-056
Specifications
About this item
Type: Primary
Antigen: LAG-3
Clonality: Recombinant
Clone: C9B7W-CP014
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse