RecombiMAb Anti-CTLA-4 Mouse Recombinant Antibody [clone: 9D9-CP006]
Catalog # 77580-048
Supplier: Bio X Cell
New Product
Specifications
- Antibody Type:Primary
- Antigen Symbol:CTLA-4
- Clonality:Recombinant
- Clone:9D9-CP006
- Host:Mouse
- Reactivity:Mouse
- Western Blot:Yes
- Size:1 mg
- Tested Applications:in vivo CTLA-4 neutralization** *Reported for the original mouse IgG2b 9D9 antibody
- Cat. No.:77580-048
Specifications
About this item
Type: Primary
Antigen: CTLA-4
Clonality: Recombinant
Clone: 9D9-CP006
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse