InVivoMAb Anti-NKG2D Rat Monoclonal Antibody [clone: CX5]
Supplier: Bio X Cell
New Product
Certificates
About this item
Type: Primary
Antigen: NKG2D
Clonality: Monoclonal
Clone: CX5
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Specifications
- Catalog No:
- 77578-896
- 77578-894
- 77578-892
- 77578-890
- 77578-898
- Antigen Symbol:
- NKG2D
- NKG2D
- NKG2D
- NKG2D
- NKG2D
- Antibody Type:
- Primary
- Primary
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- CX5
- CX5
- CX5
- CX5
- CX5
- Reactivity:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Rat
- Rat
- Flow Cytometry:
- Yes
- Yes
- Yes
- Yes
- Yes
- Tested Applications:
- in vivo NKG2D blockade, in vitro NKG2D blockade
- in vivo NKG2D blockade, in vitro NKG2D blockade
- in vivo NKG2D blockade, in vitro NKG2D blockade
- in vivo NKG2D blockade, in vitro NKG2D blockade
- in vivo NKG2D blockade, in vitro NKG2D blockade
Specifications
Frequently Bought Together