ContactUsLinkComponent
Mini Hotplate, Slimline
Mini Hotplate, Slimline
Catalog # 77578-308
Supplier:  StatLab
New Product
undefined
Mini Hotplate, Slimline
Catalog # 77578-308
Supplier:  StatLab
Supplier Number:  SI-SDH-MINI
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Mini hotplate, slimline reaches a max temperature of 75 °C
  • Cat. No.:
    77578-308
Specifications
About this item
The StatLab slimline mini hotplate has a very small footprint, making it ideal for use at the microtome. It is primarily used to remove stubborn wrinkles from tissue sections, and re-orientating embedded tissue.
  • Ultra-fast heating system for rapid warm up
  • Small footprint
  • Anti-microbial powder coat finish
  • Features a matte black heated surface to provide a visual contrast for tissue sections
It may also be used in microbiology for heat fixing bacterial smears.
The body of the mini hotplate is coated with an antimicrobial finish which inhibits growth of bacteria and viruses.