Specifications
- Capacity:66.5 L
- Dimensions:17.75L×14.75W×15.5"H
- Material:Cardboard
- Cat. No.:77587-160
Specifications
About this item
TipCycle Recycle kit mail-back program provides a simple and effective solution for recycling PP#5 nonhazardous pipette tips and racks.
- Color: Brown
- Single use
- Non sterile
- Not autoclavable
- Intended use for recycling non-hazardous PP#5 pipette tips and pipette tip racks
Place liner in box, fill with nonhazardous pipette tips and racks. Seal bag with twist tie, include waiver in box, seal box, apply prepaid freight label, then ship. Recycled plastic is converted into more sustainable, long-lasting, and affordable plastic lumber for pallets, retaining walls and landscaping products.
Complete TipCycle pipette tip recycling kit includes: box, instructions for return, a box liner and liner tie to help prevent liquid spills, decontamination waiver must be completed and inserted into recycle package, return freight label placed onto top of recycle package, then simply ship the recycle package.
TipCycle stand alone recycle kit does not include pipette tips. One case of TipCycle recycle kit contains five packs of individual recycle kits.