
3-Chloro-2-methylbenzenesulfonyl chloride 98%
Catalog # TS357080010
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:1 g
- Cat. No.:TS357080010
Specifications
Specification Test Results
- Appearance:White to light yellow Crystals
- Infrared spectrum:Conforms
- Melting point:70 to 74 °C
- Titration Argentometric:≥97.5 % (ex Cl)
Specification Test Results
About this item
:
:225.09
Melting Pt:71…73 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:80563-86-6
Formula:C₇H₆Cl₂O₂S
MDL Number:MFCD00052198