Order Entry
United States
ContactUsLinkComponent
Palladium 99.9% (trace metals basis), sponge
Irritant
Flammable
Palladium 99.9% (trace metals basis), sponge
Catalog # TS268150010
New Product
CAS Number:  
Irritant
Flammable
Palladium 99.9% (trace metals basis), sponge
Catalog # TS268150010
Supplier Number:  268150010
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 g
  • Cat. No.:
    TS268150010

Specifications

Specification Test Results

  • Appearance:
    Grey Powder
  • Total trace metal impurities:
    ≤0.1 %

Specification Test Results

About this item

 :
 :106.420 g/mol
Merck Index:12,07121
Boiling Pt:3167 °C (1013 hPa)
Melting Pt:1555 °C
EINECS:231-115-6
UN:3089
 :GHS07
 :GHS02
Storage Temperature:Ambient
CAS Number:7440-05-3
Formula:Pd
MDL Number:MFCD00011167