Order Entry
United States
ContactUsLinkComponent
Polyvinylacetate, M.W. ∼100,000
Polyvinylacetate, M.W. ∼100,000
Catalog # TS183265000
New Product
CAS Number:  
Polyvinylacetate, M.W. ∼100,000
Catalog # TS183265000
Supplier Number:  183265000
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    500 g
  • Cat. No.:
    TS183265000

Specifications

Specification Test Results

  • Appearance:
    Clear Beads
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5 %
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
Density:1.18 g/cm³
Storage Temperature:Ambient
CAS Number:9003-20-7
Formula:[CH₂CH(O₂CCH₃)]n
MDL Number:MFCD00084457