Poly(methyl methacrylate), M.W. 90000∼, fine granules secondary standard
Catalog # TS178780250
Supplier: Thermo Scientific Chemicals
New Product
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Glass bottle
- Size:25 g
- Cat. No.:TS178780250
Specifications
Specification Test Results
- Appearance:White Beads
- Infrared spectrum:Conforms
- Loss on drying:≤5 %
- Particle size:50 to 150 microns (Approximately)
Specification Test Results
About this item
:
Melting Pt:>150 °C
Flash Pt:303 °C
Storage Temperature:Ambient
CAS Number:9011-14-7
Formula:C₅H₈O₂([-CH₂C(CH₃)(CO₂CH₃)-]n)
MDL Number:MFCD00134349