Order Entry
United States
ContactUsLinkComponent
 
Wheat Germ Agglutinin (WGA), iFluor™ 488
Catalog # MSPP-100-0816
CAS Number:  
undefined
Wheat Germ Agglutinin (WGA), iFluor™ 488
Catalog # MSPP-100-0816
Supplier Number:  100-0816
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Source:
  • Species:
  • Size:
    1000 µg
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-100-0816

Specifications

About this item

Fluorescent dye conjugates for staining cell membranes of gram-positive bacteria, mammalian cells, and yeast bud scars - 1000 µg.

Wheat germ agglutinins (WGA) are lectins or carbohydrate-binding proteins that have a high affinity for N-acetyl-D-glucosamine and sialic acid present in the cell membranes. Their iFluor™ conjugates are used to stain cell membranes and fibrotic scar tissue for fluorescence imaging and analysis.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').